3,5-Dihydroxybenzaldehyd

Lieferant: Molekula

20449406-1G
MOLE20449406-1GEA 0 EUR
MOLE20449406-1G
3,5-Dihydroxybenzaldehyd
3,5-Dihydroxybenzaldehyde
Formel: C₇H₆O₃
Molecular Weight: 138,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016611
CAS-Nummer: 26153-38-8
EINECS: 247-479-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR