N-[(Benzyloxy)carbonyl]-L-tryptophan

Lieferant: Molekula

Synonyme: (S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid, N(α)-Benzyloxycarbonyl-L-tryptophan, N-Cbz-L-Tryptophan, Z-Trp-OH

20706773-25G
MOLE20706773-25GEA 71.6 EUR
MOLE20706773-25G
N-[(Benzyloxy)carbonyl]-L-tryptophan
N(α)-Cbz-L-tryptophan
Formel: C₁₉H₁₈N₂O₄
Molecular Weight: 338,4 g/mol
MDL: MFCD00065700
CAS-Nummer: 7432-21-5
EINECS: 231-074-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR