3-Amino-1H-1,2,4-triazol-5-thiol
Lieferant: Molekula
Synonyme:
3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol
20831626-25G
MOLE20831626-25GEA
0
EUR
MOLE20831626-25G
3-Amino-1H-1,2,4-triazol-5-thiol
3-Amino-1H-1,2,4-triazol-5-thiol
Formel:
C₂H₄N₄S Molecular Weight: 116,15 g/mol |
MDL:
MFCD00005231 CAS-Nummer: 16691-43-3 EINECS: 240-735-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...