5-Brom-3-indolylessigsäure
Lieferant: Molekula
Synonyme:
5-Bromindol-3-essigsäure, 5-Bromindol-3-ylessigsäure
20843123-1G
MOLE20843123-1GEA
0
EUR
MOLE20843123-1G
5-Brom-3-indolylessigsäure
5-Brom-3-indolylessigsäure
Formel:
C₁₀H₈BrNO₂ Molecular Weight: 254,08 g/mol Schmelzpunkt: 174,5 °C Dichte: 1,754 g/cm³ (20 °C) |
MDL:
MFCD00005637 CAS-Nummer: 40432-84-6 EINECS: 254-917-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...