N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10)
Lieferant: Molekula
|
Achtung
|
Synonyme:
3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate
20889868-25G
MOLE20889868-25GEA
429
EUR
MOLE20889868-25G
N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10)
N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10)
Formel:
C₁₅H₃₃NO₃S Molecular Weight: 307,4924 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036908 CAS-Nummer: 15163-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...