5-(4-Methylphenyl)-1H-tetrazol
Lieferant: Molekula
Synonyme:
5-(p-Tolyl)-1H-tetrazole
21248002-100G
MOLE21248002-100GEA
0
EUR
MOLE21248002-100G
5-(4-Methylphenyl)-1H-tetrazol
5-(4-Methylphenyl)-1H-tetrazole
Formel:
C₈H₈N₄ Molecular Weight: 160,18 g/mol |
MDL:
MFCD01318164 CAS-Nummer: 24994-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...