Cystaminiumdichlorid

Lieferant: Molekula

21267602-100G
MOLE21267602-100GEA 0 EUR
MOLE21267602-100G
Cystaminiumdichlorid
Cystaminiumdichlorid
Formel: C₄H₁₂N₂S₂·2HCl
MDL: MFCD00012905
CAS-Nummer: 56-17-7
EINECS: 200-260-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR