N-Methyl-N-nonanoyl-D-glucamin (MEGA-9)

Lieferant: Molekula

21276387-1G
MOLE21276387-1GEA 181 EUR
MOLE21276387-1G
N-Methyl-N-nonanoyl-D-glucamin (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamin (MEGA-9)
Formel: C₁₆H₃₃NO₆
Lagertemperatur: Raumtemperatur
CAS-Nummer: 85261-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR