Dodecyl-β-D-glucopyranosid

Lieferant: Molekula

Synonyme: Dodecyl-β-D-glucosid

21301716-1G
MOLE21301716-1GEA 216 EUR
MOLE21301716-1G
Dodecyl-β-D-glucopyranosid
Dodecyl-β-D-glucopyranosid
Formel: C₁₈H₃₆O₆
Molecular Weight: 348,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063298
CAS-Nummer: 59122-55-3
EINECS: 261-614-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR