3-Hydroxyzimtsäure

Lieferant: Molekula
Achtung

Synonyme: m-Hydroxycinnamic acid, 3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid

21455098-100G
MOLE21455098-100GEA 0 EUR
MOLE21455098-100G
3-Hydroxyzimtsäure
3-Hydroxyzimtsäure
Formel: C₉H₈O₃
Molecular Weight: 164,158 g/mol
MDL: MFCD00004390
CAS-Nummer: 588-30-7
EINECS: 209-615-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR