n-Dodecyl-β-D-maltosid

Lieferant: Molekula
Achtung

Synonyme: DDM

21771928-1G
MOLE21771928-1GEA 89.4 EUR
MOLE21771928-1G
n-Dodecyl-β-D-maltosid
n-Dodecyl-β-D-maltosid
Formel: C₂₄H₄₆O₁₁
Molecular Weight: 510,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00043012
CAS-Nummer: 69227-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR