Zinn(II)trifluormethansulfonat

Lieferant: Molekula

Synonyme: Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

21892641-1G
MOLE21892641-1GEA 0 EUR
MOLE21892641-1G
Zinn(II)trifluormethansulfonat
Zinn(II)trifluormethansulfonat
Formel: C₂F₆O₆S₂Sn
Molecular Weight: 416,85 g/mol
MDL: MFCD00191251
CAS-Nummer: 62086-04-8
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR