TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)
Lieferant: Molekula
Synonyme:
3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid
21958982-500G
MOLE21958982-500GEA
300
EUR
MOLE21958982-500G
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)
TAPS (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure)
Formel:
C₇H₁₇NO₆S Molecular Weight: 243,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007538 CAS-Nummer: 29915-38-6 EINECS: 249-954-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...