N,N-Dimethylglycin

Lieferant: Molekula

Synonyme: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

21966200-25G
MOLE21966200-25GEA 157 EUR
MOLE21966200-25G
N,N-Dimethylglycin
N,N-Dimethylglycin
Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
MDL: MFCD00004283
CAS-Nummer: 1118-68-9
EINECS: 214-267-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR