Sudanschwarz B
Lieferant: Molekula
Synonyme:
CI 26150, Ceres black BN, Solvent Black 3, Fat Black HB
21998092-25G
MOLE21998092-25GEA
88.1
EUR
MOLE21998092-25G
Sudanschwarz B
Sudanschwarz B
Formel:
C₂₉H₂₄N₆ Molecular Weight: 456.55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006919 CAS-Nummer: 4197-25-5 EINECS: 224-087-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...