N-Acetyl-L-methionin
Lieferant: Molekula
Synonyme:
N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH
22003320-100G
MOLE22003320-100GEA
160
EUR
MOLE22003320-100G
N-Acetyl-L-methionin
N-Acetyl-L-methionin
Formel:
C₇H₁₃NO₃S Molecular Weight: 191.2 g/mol |
MDL:
MFCD00064441 CAS-Nummer: 65-82-7 EINECS: 200-617-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...