5-Bromuridin

Lieferant: Molekula

22067520-1G
MOLE22067520-1GEA 89.4 EUR
MOLE22067520-1G
5-Bromuridin
5-Bromuridin
Formel: C₉H₁₁BrN₂O₆
Molecular Weight: 323,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006528
CAS-Nummer: 957-75-5
EINECS: 213-486-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR