N-Decanoyl-N-methyl-D-glucamin (MEGA-10)

Lieferant: Molekula

22118459-1G
MOLE22118459-1GEA 180 EUR
MOLE22118459-1G
N-Decanoyl-N-methyl-D-glucamin (MEGA-10)
N-Decanoyl-N-methyl-D-glucamin (MEGA-10)
Formel: C₁₇H₃₅NO₆
Molecular Weight: 349,46 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036801
CAS-Nummer: 85261-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR