1-Amino-2-naphthol Hydrochlorid

Lieferant: Molekula

22144792-5G
MOLE22144792-5GEA 0 EUR
MOLE22144792-5G
1-Amino-2-naphthol Hydrochlorid
1-Amino-2-naphthol Hydrochlorid
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195.65 g/mol
MDL: MFCD00012561
CAS-Nummer: 1198-27-2
EINECS: 214-830-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR