3-Amino-9-ethylcarbazol (AEC)
Lieferant: Molekula
|
Gefahr
|
Synonyme:
9-Ethylcarbazol-3-ylamin, AEC
22225310-100G
MOLE22225310-100GEA
446
EUR
MOLE22225310-100G
3-Amino-9-ethylcarbazol (AEC)
3-Amino-9-ethyl carbazole (AEC)
Formel:
C₁₄H₁₄N₂ Molecular Weight: 210,28 g/mol Schmelzpunkt: 113…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004964 CAS-Nummer: 132-32-1 EINECS: 205-057-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...