N-Cbz-Glycin (Z-Gly-OH)

Lieferant: Molekula

Synonyme: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

22586147-100G
MOLE22586147-100GEA 109 EUR
MOLE22586147-100G
N-Cbz-Glycin (Z-Gly-OH)
N-Cbz-Glycin (Z-Gly-OH)
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,2 g/mol
MDL: MFCD00002691
CAS-Nummer: 1138-80-3
EINECS: 214-516-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR