Kieselgel, Porengröße: 60 Å

Lieferant: Molekula

22768261-250G
MOLE22768261-250GEA 0 EUR
MOLE22768261-250G
Kieselgel, Porengröße: 60 Å
Silica gel
Formel: SiO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR