5-Fluoruridin
Lieferant: Molekula
Synonyme:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione
22848333-100MG
MOLE22848333-100MGEA
0
EUR
MOLE22848333-100MG
5-Fluoruridin
5-Fluoruridin
Formel:
C₉H₁₁FN₂O₆ Molecular Weight: 262,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036832 CAS-Nummer: 316-46-1 EINECS: 206-260-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...