Apigenin

Lieferant: Molekula

Synonyme: 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-on, 4',5,7-Trihydroxyflavon, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyron, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-on

22930039-25MG
MOLE22930039-25MGEA 0 EUR
MOLE22930039-25MG
Apigenin
Apigenin
Formel: C₁₅H₁₀O₅
Molecular Weight: 270,24 g/mol
MDL: MFCD00006831
CAS-Nummer: 520-36-5
EINECS: 208-292-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR