1-Diazo-2-naphthol-4-sulfonsäure

Lieferant: Molekula

23051132-100G
MOLE23051132-100GEA 194 EUR
MOLE23051132-100G
1-Diazo-2-naphthol-4-sulfonsäure
1-Diazo-2-naphthol-4-sulfonsäure
Formel: C₁₀H₆N₂O₄S
Molecular Weight: 250,23 g/mol
MDL: MFCD00012409
CAS-Nummer: 887-76-3
EINECS: 212-958-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR