5-Nitrovanillin

Lieferant: Molekula

Synonyme: 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

23063193-25G
MOLE23063193-25GEA 182 EUR
MOLE23063193-25G
5-Nitrovanillin
5-Nitrovanillin
Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
Schmelzpunkt: 175…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007118
CAS-Nummer: 6635-20-7
EINECS: 229-633-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR