Triethylphosphonoacetat

Lieferant: Molekula

23527646-100G
MOLE23527646-100GEA 0 EUR
MOLE23527646-100G
Triethylphosphonoacetat
Triethylphosphonoacetat
Formel: C₈H₁₇O₅P
MDL: MFCD00009177
CAS-Nummer: 867-13-0
EINECS: 212-757-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR