L(+)-Ribose
Lieferant: Molekula
Synonyme:
(2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose
23543542-1G
MOLE23543542-1GEA
0
EUR
MOLE23543542-1G
L(+)-Ribose
L(+)-Ribose
Formel:
C₅H₁₀O₅ Molecular Weight: 150,13 g/mol Siedepunkt: 331 °C (1013 hPa) Schmelzpunkt: 86…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167010 CAS-Nummer: 24259-59-4 EINECS: 246-110-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...