2-Chlor-4-nitrophenol

Lieferant: Molekula

23576288-25G
MOLE23576288-25GEA 0 EUR
MOLE23576288-25G
2-Chlor-4-nitrophenol
2-Chlor-4-nitrophenol
Formel: C₆H₄ClNO₃
Schmelzpunkt: 109…112 °C
MDL: MFCD00043910
CAS-Nummer: 619-08-9
EINECS: 210-578-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR