2,3,3-Trimethylindolenin

Lieferant: Molekula

23843892-25G
MOLE23843892-25GEA 0 EUR
MOLE23843892-25G
2,3,3-Trimethylindolenin
2,3,3-Trimethylindolenin
Formel: C₁₁H₁₃N
Molecular Weight: 159,23 g/mol
MDL: MFCD00005724
CAS-Nummer: 1640-39-7
EINECS: 216-685-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR