2-Methoxyphenylboronsäure

Lieferant: Molekula

Synonyme: (2-Methoxyphenyl)metaborsäure, Dihydroxy(2-methoxyphenyl)boran, 2-Methoxybenzolboronsäure

24085771-1G
MOLE24085771-1GEA 0 EUR
MOLE24085771-1G
2-Methoxyphenylboronsäure
2-Methoxyphenylboronsäure
Formel: C₇H₉BO₃
Molecular Weight: 151,96 g/mol
Schmelzpunkt: 165 °C
MDL: MFCD00236047
CAS-Nummer: 5720-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR