3-Fluorstyrol

Lieferant: Molekula

24155756-5G
MOLE24155756-5GEA 0 EUR
MOLE24155756-5G
3-Fluorstyrol
3-Fluorstyrol
Formel: C₈H₇F
Molecular Weight: 122,14 g/mol
Siedepunkt: 30 °C (1013 hPa)
Dichte: 1,025 g/cm³ (20 °C)
MDL: MFCD00000341
CAS-Nummer: 350-51-6
EINECS: 206-504-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR