2-Methyl-3-pyridinol

Lieferant: Molekula

Synonyme: 3-Hydroxy-2-methylpyridin

24215178-5G
MOLE24215178-5GEA 0 EUR
MOLE24215178-5G
2-Methyl-3-pyridinol
2-Methylpyridin-3-ol
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00082538
CAS-Nummer: 1121-25-1
EINECS: 214-327-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR