X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)

Lieferant: Molekula

Synonyme: Cyclohexylammoniumsalz der 5-Brom-4-chlor-3-indolyl-β-D-glucuronsäure, X-Glucuro CHAsalz, Cyclohexylammoniumsalz der (2S,3S,4S,5R)-6-[(5-Brom-4-chlor-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxan-2-carbonsäure

24519211-100MG
MOLE24519211-100MGEA 0 EUR
MOLE24519211-100MG
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)
Formel: C₂₀H₂₆BrClN₂O₇
Molecular Weight: 521,79 g/mol
Schmelzpunkt: 230 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00058543
CAS-Nummer: 114162-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR