4-Methylveratrol

Lieferant: Molekula
Achtung

Synonyme: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

24539542-100G
MOLE24539542-100GEA 0 EUR
MOLE24539542-100G
4-Methylveratrol
4-Methylveratrol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016651
CAS-Nummer: 494-99-5
EINECS: 207-796-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR