1-Naphthylphosphat Dinatriumsalz Monohydrat

Lieferant: Molekula

Synonyme: 1-Naphthyl phosphate, disodium salt hydrate

24815696-5G
MOLE24815696-5GEA 493 EUR
MOLE24815696-5G
1-Naphthylphosphat Dinatriumsalz Monohydrat
1-Naphthylphosphat Dinatriumsalz Monohydrat
Formel: C₁₀H₇Na₂O₄P·1H₂O
Molecular Weight: 268,12 g/mol (anhy)
MDL: MFCD00150614
CAS-Nummer: 207569-06-0
EINECS: 218-564-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR