2-Amino-5-chlor-2'-fluorbenzophenon
Lieferant: Molekula
Synonyme:
(2-Amino-5-chlorophenyl)(2-fluorophenyl)methanone, 2-Amino-5-chloro-2′-fluorobenzophenone
24826290-5G
MOLE24826290-5GEA
0
EUR
MOLE24826290-5G
2-Amino-5-chlor-2'-fluorbenzophenon
2-Amino-5-chlor-2'-fluorbenzophenon
Formel:
C₁₃H₉ClFNO Molecular Weight: 249,67 g/mol Siedepunkt: 364 °C (1013 hPa) Schmelzpunkt: 94…95 °C |
MDL:
MFCD00038381 CAS-Nummer: 784-38-3 EINECS: 212-316-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...