(R)-(-)-Thiazolidin-4-carbonsäure
Lieferant: Molekula
Synonyme:
H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid
24896910-25G
MOLE24896910-25GEA
0
EUR
MOLE24896910-25G
(R)-(-)-Thiazolidin-4-carbonsäure
(R)-(-)-Thiazolidin-4-carbonsäure
Formel:
C₄H₇NO₂S Molecular Weight: 133,17 g/mol |
MDL:
MFCD00005212 CAS-Nummer: 34592-47-7 EINECS: 252-106-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...