Naphthol AS-acetat

Lieferant: Molekula

Synonyme: 2-(N-Phenylcarbamoyl)-3-naphthylacetat

25134487-5G
MOLE25134487-5GEA 219 EUR
MOLE25134487-5G
Naphthol AS-acetat
Naphthol AS-acetat
Formel: C₁₉H₁₅NO₃
Molecular Weight: 305,33 g/mol
MDL: MFCD00004095
CAS-Nummer: 1163-67-3
EINECS: 214-608-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR