2,4,6-Trichlorbenzonitril

Lieferant: Molekula

25180755-5G
MOLE25180755-5GEA 0 EUR
MOLE25180755-5G
2,4,6-Trichlorbenzonitril
2,4,6-Trichlorbenzonitril
Formel: C₇H₂Cl₃N
Molecular Weight: 206,46 g/mol
MDL: MFCD00052711
CAS-Nummer: 6575-05-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR