5-Chlorindol

Lieferant: Molekula

Synonyme: 5-Chloro-1H-indole

25238892-5G
MOLE25238892-5GEA 0 EUR
MOLE25238892-5G
5-Chlorindol
5-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
Schmelzpunkt: 71…72 °C
MDL: MFCD00005672
CAS-Nummer: 17422-32-1
EINECS: 241-448-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR