2,5-Diaminopyridin Dihydrochlorid

Lieferant: Molekula

25440537-25G
MOLE25440537-25GEA 0 EUR
MOLE25440537-25G
2,5-Diaminopyridin Dihydrochlorid
2,5-Diaminopyridin Dihydrochlorid
Formel: C₅H₉Cl₂N₃
Molecular Weight: 182.05 g/mol
Schmelzpunkt: 264
MDL: MFCD00040818
CAS-Nummer: 26878-35-3
EINECS: 248-083-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR