2-Fluorstyrol
Lieferant: Molekula
Synonyme:
1-Fluoro-2-vinylbenzene
25473627-5G
MOLE25473627-5GEA
0
EUR
MOLE25473627-5G
2-Fluorstyrol
2-Fluorstyrol
Formel:
C₈H₇F Molecular Weight: 122,14 g/mol Siedepunkt: 94 °C (0,5 mmHg) Dichte: 1,03 g/cm³ (20 °C) |
MDL:
MFCD00013550 CAS-Nummer: 394-46-7 EINECS: 206-896-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...