1,4-Diaminobutan Dihydrochlorid

Lieferant: Molekula

Synonyme: 1,4-Butandiamin Dihydrochlorid, Tetramethylendiammoniumdichlorid, Tetramethylendiamin Dihydrochlorid, Putrescin Dihydrochlorid

25523156-25G
MOLE25523156-25GEA 93.4 EUR
MOLE25523156-25G
1,4-Diaminobutan Dihydrochlorid
1,4-Diaminobutan Dihydrochlorid
Formel: C₄H₁₄Cl₂N₂
Molecular Weight: 161,07 g/mol
Dichte: 1,31 g/cm³ (20 °C)
MDL: MFCD00012526
CAS-Nummer: 333-93-7
EINECS: 206-375-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR