Triethanolamin Hydrochlorid
Lieferant: Molekula
25559545-100G
MOLE25559545-100GEA
71.6
EUR
MOLE25559545-100G
Triethanolamin Hydrochlorid
Triethanolamin Hydrochlorid
Formel:
C₆H₁₆ClNO₃ Molecular Weight: 185,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012596 CAS-Nummer: 637-39-8 EINECS: 211-284-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...