4-Nitroindol

Lieferant: Molekula

Synonyme: 4-Nitro-1H-indole

25699714-1G
MOLE25699714-1GEA 0 EUR
MOLE25699714-1G
4-Nitroindol
4-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 206…209 °C
MDL: MFCD00010056
CAS-Nummer: 4769-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR