3,4-Dimethoxybenzylalkohol

Lieferant: Molekula

Synonyme: Veratrylalkohol, (3,4-Dimethoxyphenyl)methanol

25751471-100G
MOLE25751471-100GEA 0 EUR
MOLE25751471-100G
3,4-Dimethoxybenzylalkohol
3,4-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
MDL: MFCD00004638
CAS-Nummer: 93-03-8
EINECS: 202-212-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR