2-Cyanbenzaldehyd

Lieferant: Molekula

Synonyme: 2-Formylbenzonitrile

25881995-5G
MOLE25881995-5GEA 0 EUR
MOLE25881995-5G
2-Cyanbenzaldehyd
2-Cyanbenzaldehyd
Formel: C₈H₅NO
Molecular Weight: 131,13 g/mol
MDL: MFCD00017503
CAS-Nummer: 7468-67-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR