2-Bromphenetol

Lieferant: Molekula

Synonyme: 1-Brom-2-ethoxybenzol, (2-Bromphenyl)ethylether

25962068-5G
MOLE25962068-5GEA 89.7 EUR
MOLE25962068-5G
2-Bromphenetol
2-Bromphenetol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00061092
CAS-Nummer: 583-19-7
EINECS: 209-501-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR