4-Aminoindol

Lieferant: Molekula

Synonyme: Indol-4-ylamin, 4-Indolamin, 4-Amino-1H-indol

25980595-100MG
MOLE25980595-100MGEA 0 EUR
MOLE25980595-100MG
4-Aminoindol
4-Aminoindol
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
Schmelzpunkt: 108 °C
MDL: MFCD01076559
CAS-Nummer: 5192-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR